Tisk…

4-Bromo-2-chloro-6-(trifluoromethoxy)aniline

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Amino-5-bromo-3-chloro-α,α,α-trifluoroanisole

PC48298-1G PC48298-250MG PC48298-5G
APOSPC48298-1GEA 622 CZK
APOSPC48298-1G APOSPC48298-250MG APOSPC48298-5G
4-Bromo-2-chloro-6-(trifluoromethoxy)aniline
4-Bromo-2-chloro-6-(trifluoromethoxy)aniline
Vzorec: C₇H₄BrClF₃NO
MW: 290,465 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08236694
CAS číslo: 885266-98-8
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR