3-Bromo-4-chloro-2-fluorobenzoic acid 95%
Dodavatel: Apollo Scientific
PC48027-1G
PC48027-250MG
APOSPC48027-1GEA
19010
CZK
APOSPC48027-1G
APOSPC48027-250MG
3-Bromo-4-chloro-2-fluorobenzoic acid 95%
3-Bromo-4-chloro-2-fluorobenzoic acid
Vzorec:
C₇H₃BrClFO₂ MW: 253,453 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28398041 CAS číslo: 1677706-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...