3-Bromo-4-chloro-2-fluorobenzyl bromide 95%
Dodavatel: Apollo Scientific
Synonyms:
3-Bromo-1-(bromomethyl)-4-chloro-2-fluorobenzene
PC48004-1G
PC48004-250MG
APOSPC48004-1GEA
22320
CZK
APOSPC48004-1G
APOSPC48004-250MG
3-Bromo-4-chloro-2-fluorobenzyl bromide 95%
3-Bromo-4-chloro-2-fluorobenzyl bromide
Vzorec:
C₇H₄Br₂ClF MW: 302,366 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31915844 CAS číslo: 2092617-72-4 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...