Methyl 5-bromo-2,3,4-trifluorobenzoate 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC48003-1G
PC48003-250MG
PC48003-5G
APOSPC48003-1GEA
827
CZK
APOSPC48003-1G
APOSPC48003-250MG
APOSPC48003-5G
Methyl 5-bromo-2,3,4-trifluorobenzoate 95%
Methyl 5-bromo-2,3,4-trifluorobenzoate
Vzorec:
C₈H₄BrF₃O₂ MW: 269,015 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12911527 CAS číslo: 530145-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...