Tisk…

3-Bromo-4-chloro-2-fluorobenzonitrile 95%

Dodavatel: Apollo Scientific

PC47693-1G PC47693-250MG
APOSPC47693-1GEA 19010 CZK
APOSPC47693-1G APOSPC47693-250MG
3-Bromo-4-chloro-2-fluorobenzonitrile 95%
3-Bromo-4-chloro-2-fluorobenzonitrile
Vzorec: C₇H₂BrClFN
Storage Temperature: Ambient
MDL Number: MFCD31924547
CAS číslo: 2092624-38-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR