Tisk…

2-Bromo-3,5-difluoroaniline 95+%

Dodavatel: Apollo Scientific
Varování

PC47688-1G PC47688-5G PC47688-10G
APOSPC47688-1GEA 622 CZK
APOSPC47688-1G APOSPC47688-5G APOSPC47688-10G
2-Bromo-3,5-difluoroaniline 95+%
2-Bromo-3,5-difluoroaniline
Vzorec: C₆H₄BrF₂N
MW: 208.003 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16661040
CAS číslo: 500357-40-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR