Tisk…

2,4-Difluoro-3-(trifluoromethyl)aniline 95%

Dodavatel: Apollo Scientific
Varování

PC47684-1G PC47684-250MG
APOSPC47684-1GEA 15150 CZK
APOSPC47684-1G APOSPC47684-250MG
2,4-Difluoro-3-(trifluoromethyl)aniline 95%
2,4-Difluoro-3-(trifluoromethyl)aniline
Vzorec: C₇H₄F₅N
Storage Temperature: Ambient
MDL Number: MFCD09261074
CAS číslo: 123973-26-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR