Magnesium nonafluorobutane-1-sulphonate 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC4649-5G
PC4649-1G
APOSPC4649-5GEA
11540
CZK
APOSPC4649-5G
APOSPC4649-1G
Magnesium nonafluorobutane-1-sulphonate 97%
Magnesium nonafluorobutane-1-sulphonate
Vzorec:
2(C₄F₉O₃S)·Mg Storage Temperature: Ambient |
MDL Number:
MFCD03095412 CAS číslo: 507453-86-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...