1-(2-Chloroethyl)-3-(3,5-difluorophenyl)urea
Dodavatel: Apollo Scientific
|
Varování
|
PC46363-1G
PC46363-250MG
APOSPC46363-1GEA
744
CZK
APOSPC46363-1G
APOSPC46363-250MG
1-(2-Chloroethyl)-3-(3,5-difluorophenyl)urea
1-(2-Chloroethyl)-3-(3,5-difluorophenyl)urea
Vzorec:
C₉H₉ClF₂N₂O MW: 234.63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31372409 CAS číslo: 2169031-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...