Tisk…

1-(2-Chloroethyl)-3-(3-fluorophenyl)urea

Dodavatel: Apollo Scientific
Varování

PC46359-1G PC46359-250MG
APOSPC46359-1GEA 744 CZK
APOSPC46359-1G APOSPC46359-250MG
1-(2-Chloroethyl)-3-(3-fluorophenyl)urea
1-(2-Chloroethyl)-3-(3-fluorophenyl)urea
Vzorec: C₉H₁₀ClFN₂O
MW: 216,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14586691
CAS číslo: 146257-15-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR