2-Chloro-3,5-difluorobenzonitrile 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC46352-1G
PC46352-250MG
PC46352-5G
APOSPC46352-1GEA
1410
CZK
APOSPC46352-1G
APOSPC46352-250MG
APOSPC46352-5G
2-Chloro-3,5-difluorobenzonitrile 95%
2-Chloro-3,5-difluorobenzonitrile
Vzorec:
C₇H₂ClF₂N Storage Temperature: Ambient |
MDL Number:
MFCD18415560 CAS číslo: 1261472-31-4 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...