Ethyl N-[4-(trifluoromethyl)pyrimid-2-yl]piperidine-4-carboxylate
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 1-(4-(trifluoromethyl)pyrimidin-2-yl)piperidine-4-carboxylate
PC4631-10G
PC4631-1G
PC4631-5G
APOSPC4631-10GEA
28440
CZK
APOSPC4631-10G
APOSPC4631-1G
APOSPC4631-5G
Ethyl N-[4-(trifluoromethyl)pyrimid-2-yl]piperidine-4-carboxylate
Ethyl N-[4-(trifluoromethyl)pyrimid-2-yl]piperidine-4-carboxylate
Vzorec:
C₁₃H₁₆F₃N₃O₂ MW: 303,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00661832 CAS číslo: 215654-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...