4-Amino-3-methoxybenzotrifluoride 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Amino-5-(trifluoromethyl)anisole, 2-Methoxy-4-(trifluoromethyl)aniline
PC4598-10G
PC4598-5G
PC4598-1G
APOSPC4598-10GEA
14120
CZK
APOSPC4598-10G
APOSPC4598-5G
APOSPC4598-1G
4-Amino-3-methoxybenzotrifluoride 98%
4-Amino-3-methoxybenzotrifluoride
Vzorec:
C₈H₈F₃NO MW: 191,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115973 CAS číslo: 158727-56-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...