Tisk…

Ethyl 2,2-dimethyl-4,4,4-trifluorobutanoate

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 2,2-dimethyl-4,4,4-trifluorobutyrate

PC4550-1G
APOSPC4550-1GEA 7410 CZK
APOSPC4550-1G
Ethyl 2,2-dimethyl-4,4,4-trifluorobutanoate
Ethyl 2,2-dimethyl-4,4,4-trifluorobutanoate
Vzorec: C₈H₁₃F₃O₂
MW: 198,183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784244
CAS číslo: 885275-92-3
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR