6-Amino-2,2,3,3-tetrafluoro-1,4-benzodioxane
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2,2,3,3-Tetrafluoro-2,3-dihydro-1,4-benzodioxin-6-amine, 6-Amino-2,2,3,3-tetrafluoro-2,3-dihydro-1,4-benzodioxine
PC4536-1G
PC4536-5G
PC4536-25G
APOSPC4536-1GEA
5200
CZK
APOSPC4536-1G
APOSPC4536-5G
APOSPC4536-25G
6-Amino-2,2,3,3-tetrafluoro-1,4-benzodioxane
6-Amino-2,2,3,3-tetrafluoro-1,4-benzodioxane
Vzorec:
C₈H₅F₄NO₂ MW: 223,124 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00190146 CAS číslo: 89586-07-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...