Tisk…

1,1,1,4,4,4-Hexafluoro-2,3-bis(4-bromophenoxy)-2-butene 97%

Dodavatel: Apollo Scientific
Varování

PC450574-1G PC450574-250MG
APOSPC450574-1GEA 9720 CZK
APOSPC450574-1G APOSPC450574-250MG
1,1,1,4,4,4-Hexafluoro-2,3-bis(4-bromophenoxy)-2-butene 97%
1,1,1,4,4,4-Hexafluoro-2,3-bis(4-bromophenoxy)-2-butene
Vzorec: C₁₆H₈Br₂F₆O₂
MW: 506,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30062857
CAS číslo: 1980781-18-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR