Tisk…

(Z)-(1,2-Difluorovinyl)trimethylsilane 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC450572-1G PC450572-250MG
APOSPC450572-1GEA 24400 CZK
APOSPC450572-1G APOSPC450572-250MG
(Z)-(1,2-Difluorovinyl)trimethylsilane 97%
(Z)-(1,2-Difluorovinyl)trimethylsilane
Vzorec: C₅H₁₀F₂Si
MW: 136.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30062987
CAS číslo: 17757-16-3
UN: 1993
ADR: 3 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR