Tisk…

1,1,1,2,2-Pentafluoropentane

Dodavatel: Apollo Scientific
Nebezpečí

PC450571-1G PC450571-250MG PC450571-5G
APOSPC450571-1GEA 4140 CZK
APOSPC450571-1G APOSPC450571-250MG APOSPC450571-5G
1,1,1,2,2-Pentafluoropentane
1,1,1,2,2-Pentafluoropentane
Vzorec: C₅H₇F₅
MW: 162,101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28135687
CAS číslo: 154381-43-8
UN: 1993
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR