Tisk…

4-(Fluoromethyl)-1,3-dioxolan-2-one

Dodavatel: Apollo Scientific
Varování

PC450562-1G PC450562-250MG
APOSPC450562-1GEA 19640 CZK
APOSPC450562-1G APOSPC450562-250MG
4-(Fluoromethyl)-1,3-dioxolan-2-one
4-(Fluoromethyl)-1,3-dioxolan-2-one
Vzorec: C₄H₅FO₃
MW: 120,079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22577168
CAS číslo: 127213-73-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR