Tisk…

(4R)-4-Phenyl-2-(difluoromethyl)oxazolidine 95%

Dodavatel: Apollo Scientific
Varování

PC450560-1G PC450560-250MG
APOSPC450560-1GEA 33730 CZK
APOSPC450560-1G APOSPC450560-250MG
(4R)-4-Phenyl-2-(difluoromethyl)oxazolidine 95%
(4R)-4-Phenyl-2-(difluoromethyl)oxazolidine
Vzorec: C₁₀H₁₁F₂NO
MW: 199,197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28137321
CAS číslo: 1902254-13-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR