Tisk…

3-Bromo-1,1,1,2,2-pentafluoropropane

Dodavatel: Apollo Scientific
Varování

PC450558-1G PC450558-5G
APOSPC450558-1GEA 5630 CZK
APOSPC450558-1G APOSPC450558-5G
3-Bromo-1,1,1,2,2-pentafluoropropane
3-Bromo-1,1,1,2,2-pentafluoropropane
Vzorec: C₃H₂BrF₅
MW: 212,944 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00233030
CAS číslo: 422-01-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR