Buten-1-ylsulphur pentafluoride
Dodavatel: Apollo Scientific
|
Varování
|
PC450557-1G
PC450557-250MG
APOSPC450557-1GEA
19680
CZK
APOSPC450557-1G
APOSPC450557-250MG
Buten-1-ylsulphur pentafluoride
Buten-1-ylsulphur pentafluoride
Vzorec:
C₄H₇F₅S MW: 182,155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31561017 CAS číslo: 2149605-96-7 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...