Tisk…

Trifluoromethanesulphonylethane

Dodavatel: Apollo Scientific
Varování

PC450484-1G PC450484-250MG
APOSPC450484-1GEA 13530 CZK
APOSPC450484-1G APOSPC450484-250MG
Trifluoromethanesulphonylethane
Trifluoromethanesulphonylethane
Vzorec: C₃H₅F₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD28137331
CAS číslo: 13003-57-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR