Tisk…

Ethyl hexafluoroisopropyl carbonate

Dodavatel: Apollo Scientific

PC450418-25G PC450418-250MG PC450418-1G PC450418-5G
APOSPC450418-25GEA 43640 CZK
APOSPC450418-25G APOSPC450418-250MG APOSPC450418-1G APOSPC450418-5G
Ethyl hexafluoroisopropyl carbonate
Ethyl hexafluoroisopropyl carbonate
Vzorec: C₆H₆F₆O₃
MW: 240,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28334394
CAS číslo: 18925-64-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR