4-Amino-3-bromo-5-(trifluoromethoxy)benzoic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC45031-1G
PC45031-250MG
PC45031-5G
APOSPC45031-1GEA
1660
CZK
APOSPC45031-1G
APOSPC45031-250MG
APOSPC45031-5G
4-Amino-3-bromo-5-(trifluoromethoxy)benzoic acid 95%
4-Amino-3-bromo-5-(trifluoromethoxy)benzoic acid
Vzorec:
C₈H₅BrF₃NO₃ MW: 300.029 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15527568 CAS číslo: 453565-89-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...