Bis(hexafluoroisopropyl) carbonate
Dodavatel: Apollo Scientific
PC450303-25G
PC450303-5G
APOSPC450303-25GEA
53890
CZK
APOSPC450303-25G
APOSPC450303-5G
Bis(hexafluoroisopropyl) carbonate
Bis(hexafluoroisopropyl) carbonate
Vzorec:
C₇H₂F₁₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD28334297 CAS číslo: 18925-66-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...