Tisk…

2,2-Difluoroethyl 2,2,2-trifluoroethyl carbonate

Dodavatel: Apollo Scientific
Varování

PC450293-1G PC450293-5G
APOSPC450293-1GEA 5630 CZK
APOSPC450293-1G APOSPC450293-5G
2,2-Difluoroethyl 2,2,2-trifluoroethyl carbonate
2,2-Difluoroethyl 2,2,2-trifluoroethyl carbonate
Vzorec: C₅H₅F₅O₃
MW: 208,083 g/mol
Bod varu: 77…78 °C (75 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD28334288
CAS číslo: 1980085-24-2
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR