Tisk…

2,2-Difluoroethyl methyl carbonate

Dodavatel: Apollo Scientific
Varování

PC450291-1G PC450291-5G
APOSPC450291-1GEA 4300 CZK
APOSPC450291-1G APOSPC450291-5G
2,2-Difluoroethyl methyl carbonate
2,2-Difluoroethyl methyl carbonate
Vzorec: C₄H₆F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28054293
CAS číslo: 916678-13-2
UN: 1993
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR