Tisk…

Perfluorotetrahydro-2H-pyran-2-carboxylic acid

Dodavatel: Apollo Scientific
Nebezpečí

PC450285-1G PC450285-5G
APOSPC450285-1GEA 3150 CZK
APOSPC450285-1G APOSPC450285-5G
Perfluorotetrahydro-2H-pyran-2-carboxylic acid
Perfluorotetrahydro-2H-pyran-2-carboxylic acid
Vzorec: C₆HF₉O₃
MW: 292,056 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28054326
CAS číslo: 2248063-91-2
UN: 1760
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR