Tisk…

2,3-Dibromo-3-chloro-1,1,1-trifluoropropane

Dodavatel: Apollo Scientific

PC450282-1G PC450282-5G
APOSPC450282-1GEA 5720 CZK
APOSPC450282-1G APOSPC450282-5G
2,3-Dibromo-3-chloro-1,1,1-trifluoropropane
2,3-Dibromo-3-chloro-1,1,1-trifluoropropane
Vzorec: C₃H₂Br₂ClF₃
Storage Temperature: Ambient
MDL Number: MFCD18074328
CAS číslo: 1309602-11-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR