Tisk…

Ethyl dichlorofluoroacetate 96%

Dodavatel: Apollo Scientific
Nebezpečí

PC450268-25G PC450268-5G PC450268-1G
APOSPC450268-25GEA 0 CZK
APOSPC450268-25G APOSPC450268-5G APOSPC450268-1G
Ethyl dichlorofluoroacetate 96%
Ethyl dichlorofluoroacetate
Vzorec: C₄H₅Cl₂FO₂
Bod varu: 130…132 °C
Hustota: 1,307 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013664
CAS číslo: 383-61-9
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR