Tisk…

2,4-Difluoro-1,3,5-trimethylbenzene 95%

Dodavatel: Apollo Scientific

PC450262-1G PC450262-5G
APOSPC450262-1GEA 3110 CZK
APOSPC450262-1G APOSPC450262-5G
2,4-Difluoro-1,3,5-trimethylbenzene 95%
2,4-Difluoro-1,3,5-trimethylbenzene
Vzorec: C₉H₁₀F₂
Storage Temperature: Ambient
MDL Number: MFCD18074339
CAS číslo: 392-61-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR