Tisk…

2,2-Difluoroethyl difluoroacetate

Dodavatel: Apollo Scientific

PC450252-25G PC450252-5G
APOSPC450252-25GEA 31640 CZK
APOSPC450252-25G APOSPC450252-5G
2,2-Difluoroethyl difluoroacetate
2,2-Difluoroethyl difluoroacetate
Vzorec: C₄H₄F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD18074351
CAS číslo: 1300698-74-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR