Nonafluoropentanamide 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC450244-1G
PC450244-25G
PC450244-5G
APOSPC450244-1GEA
870
CZK
APOSPC450244-1G
APOSPC450244-25G
APOSPC450244-5G
Nonafluoropentanamide 97%
Nonafluoropentanamide
Vzorec:
C₅H₂F₉NO Bod varu: 180…182 °C Bod tání: 135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00451833 CAS číslo: 13485-61-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...