Tisk…

5,5,6,6,7,7,8,8,8-Nonafluoro-2-octanone 97%

Dodavatel: Apollo Scientific
Varování

PC450220-1G PC450220-5G
APOSPC450220-1GEA 4370 CZK
APOSPC450220-1G APOSPC450220-5G
5,5,6,6,7,7,8,8,8-Nonafluoro-2-octanone 97%
5,5,6,6,7,7,8,8,8-Nonafluoro-2-octanone
Vzorec: C₈H₇F₉O
Storage Temperature: Ambient
MDL Number: MFCD18072183
CAS číslo: 140834-64-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR