Tisk…

Sodium pentafluorobenzenesulphonate

Dodavatel: Apollo Scientific
Varování

PC450218-25G PC450218-5G
APOSPC450218-25GEA 31610 CZK
APOSPC450218-25G APOSPC450218-5G
Sodium pentafluorobenzenesulphonate
Sodium pentafluorobenzenesulphonate
Vzorec: C₆F₅O₃S·Na
Storage Temperature: Ambient
MDL Number: MFCD04038122
CAS číslo: 120193-44-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR