Tisk…

2-(2,6-Difluorophenyl)-1H-benzimidazole

Dodavatel: Apollo Scientific

PC450216-1G PC450216-5G
APOSPC450216-1GEA 5200 CZK
APOSPC450216-1G APOSPC450216-5G
2-(2,6-Difluorophenyl)-1H-benzimidazole
2-(2,6-Difluorophenyl)-1H-benzimidazole
Vzorec: C₁₃H₈F₂N₂
MW: 230,213 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06490681
CAS číslo: 164593-05-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR