2-(Fluorosulphonyl)difluoroacetyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC450201-1G
PC450201-250MG
PC450201-5G
APOSPC450201-1GEA
5000
CZK
APOSPC450201-1G
APOSPC450201-250MG
APOSPC450201-5G
2-(Fluorosulphonyl)difluoroacetyl chloride
2-(Fluorosulphonyl)difluoroacetyl chloride
Vzorec:
C₂ClF₃O₃S Bod varu: 67…68 °C Hustota: 1,6419 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD22054904 CAS číslo: 1648-30-2 UN: 2927 ADR: 6.1+8,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...