4-(Difluoromethoxy)-3-methoxybenzaldehyde oxime
Dodavatel: Apollo Scientific
|
Varování
|
PC450199-1G
PC450199-5G
APOSPC450199-1GEA
3110
CZK
APOSPC450199-1G
APOSPC450199-5G
4-(Difluoromethoxy)-3-methoxybenzaldehyde oxime
4-(Difluoromethoxy)-3-methoxybenzaldehyde oxime
Vzorec:
C₉H₉F₂NO₃ MW: 217.169 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01124384 CAS číslo: 733796-08-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...