Tisk…

(2,2-Difluoroethoxy)benzene

Dodavatel: Apollo Scientific
Varování

PC450195-1G PC450195-250MG PC450195-5G
APOSPC450195-1GEA 8070 CZK
APOSPC450195-1G APOSPC450195-250MG APOSPC450195-5G
(2,2-Difluoroethoxy)benzene
(2,2-Difluoroethoxy)benzene
Vzorec: C₈H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD19442073
CAS číslo: 1309602-18-3
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR