Tisk…

2,2,3,3,3-Pentafluoropropyl butyrate

Dodavatel: Apollo Scientific
Varování

PC450190-1G PC450190-5G
APOSPC450190-1GEA 2820 CZK
APOSPC450190-1G APOSPC450190-5G
2,2,3,3,3-Pentafluoropropyl butyrate
2,2,3,3,3-Pentafluoropropyl butyrate
Vzorec: C₇H₉F₅O₂
Storage Temperature: Ambient
MDL Number: MFCD22123979
CAS číslo: 950191-45-4
UN: 3272
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR