Tisk…

Bis(trifluoromethanesulphonyl)methane 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Bis((trifluoromethyl)sulphonyl)methane, Bis(trifluoromethylsulphonyl)methane

PC450189-1G PC450189-250MG PC450189-5G
APOSPC450189-1GEA 7010 CZK
APOSPC450189-1G APOSPC450189-250MG APOSPC450189-5G
Bis(trifluoromethanesulphonyl)methane 97%
Bis(trifluoromethanesulphonyl)methane
Vzorec: C₃H₂F₆O₄S₂
MW: 280,166 g/mol
Bod varu: 191 °C (745 mm Hg)
Bod tání: 32…36 °C
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01861122
CAS číslo: 428-76-2
EINECS: 207-052-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR