1,1,2,3,3,3-Hexafluoropropyl 1H,1H-heptafluorobutyl ether 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC450184-1G
PC450184-5G
APOSPC450184-1GEA
11540
CZK
APOSPC450184-1G
APOSPC450184-5G
1,1,2,3,3,3-Hexafluoropropyl 1H,1H-heptafluorobutyl ether 95%
1,1,2,3,3,3-Hexafluoropropyl 1H,1H-heptafluorobutyl ether
Vzorec:
C₇H₃F₁₃O MW: 350,077 g/mol Bod varu: 105 °C Hustota: 1,6292 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD22123962 CAS číslo: 1184-97-0 UN: 3271 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...