Tisk…

(1,2-Dibromo-2,2-difluoroethyl)benzene

Dodavatel: Apollo Scientific

PC450183-1G PC450183-250MG PC450183-5G
APOSPC450183-1GEA 8440 CZK
APOSPC450183-1G APOSPC450183-250MG APOSPC450183-5G
(1,2-Dibromo-2,2-difluoroethyl)benzene
(1,2-Dibromo-2,2-difluoroethyl)benzene
Vzorec: C₈H₆Br₂F₂
MW: 299,938 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23134586
CAS číslo: 384-63-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR