1,1,1,5,5,5-Hexafluoropentane
Dodavatel: Apollo Scientific
PC450178-1G
PC450178-5G
APOSPC450178-1GEA
8100
CZK
APOSPC450178-1G
APOSPC450178-5G
1,1,1,5,5,5-Hexafluoropentane
1,1,1,5,5,5-Hexafluoropentane
Vzorec:
C₅H₆F₆ MW: 180,092 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18074322 CAS číslo: 66711-82-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...