Tisk…

1H,1H,2H,2H-Perfluoropentyltrimethoxysilane

Dodavatel: Apollo Scientific
Varování

Synonyms: (3,3,4,4,5,5,5-Heptafluoropentyl)trimethoxysilane

PC450175-25G PC450175-1G PC450175-5G
APOSPC450175-25GEA 0 CZK
APOSPC450175-25G APOSPC450175-1G APOSPC450175-5G
1H,1H,2H,2H-Perfluoropentyltrimethoxysilane
1H,1H,2H,2H-Perfluoropentyltrimethoxysilane
Vzorec: C₈H₁₃F₇O₃Si
MW: 318.261 g/mol
Bod varu: 88…89 °C (70 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD18074425
CAS číslo: 109134-39-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR