2,4,6-Tris(perfluoropentyl)-s-triazine
Dodavatel: Apollo Scientific
PC450174-1G
PC450174-250MG
APOSPC450174-1GEA
9330
CZK
APOSPC450174-1G
APOSPC450174-250MG
2,4,6-Tris(perfluoropentyl)-s-triazine
2,4,6-Tris(perfluoropentyl)-s-triazine
Vzorec:
C₁₈F₃₃N₃ MW: 885,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18074371 CAS číslo: 23790-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...