Tisk…

5-Chloro-2-(trifluoromethyl)-1H-benzimidazole

Dodavatel: Apollo Scientific

PC450168-1G PC450168-5G
APOSPC450168-1GEA 6750 CZK
APOSPC450168-1G APOSPC450168-5G
5-Chloro-2-(trifluoromethyl)-1H-benzimidazole
5-Chloro-2-(trifluoromethyl)-1H-benzimidazole
Vzorec: C₈H₄ClF₃N₂
Storage Temperature: Ambient
MDL Number: MFCD00808236
CAS číslo: 656-49-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR