Tisk…

Methyl 2,3,3,3-tetrafluoro-2-(fluorosulphonyl)propionate

Dodavatel: Apollo Scientific

PC450166-5G
APOSPC450166-5GEA 11540 CZK
APOSPC450166-5G
Methyl 2,3,3,3-tetrafluoro-2-(fluorosulphonyl)propionate
Methyl 2,3,3,3-tetrafluoro-2-(fluorosulphonyl)propionate
Vzorec: C₄H₃F₅O₄S
Storage Temperature: Ambient
MDL Number: MFCD08753168
CAS číslo: 663-78-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR