trans-1H,2H-Octafluorocyclopentane
Dodavatel: Apollo Scientific
|
Varování
|
PC450158-1G
PC450158-5G
APOSPC450158-1GEA
0
CZK
APOSPC450158-1G
APOSPC450158-5G
trans-1H,2H-Octafluorocyclopentane
trans-1H,2H-Octafluorocyclopentane
Vzorec:
C₅H₂F₈ Bod varu: 51 °C Storage Temperature: Ambient |
MDL Number:
MFCD23704311 CAS číslo: 158389-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...